Assay Details
| Gene Symbol: TPPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692138.3 | XP_005692195.1 | 3 - 4 | 493 | 76 | |
| XM_005692139.3 | XP_005692196.1 | 3 - 4 | 553 | 76 | ||
| GenBank mRNA | JO505607.2 | - | 3 - 4 | 467 | 76 | |
| JT723651.1 | - | 2 - 3 | 511 | 76 | ||
More Information
Set Membership: |
|

TPPP3
Genomic Map