Assay Details
| Gene Symbol: PDCD2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062296.1 | XP_017917785.1 | 3 - 4 | 377 | 61 | |
| GenBank mRNA | JO610229.2 | - | 3 - 4 | 326 | 61 | |
More Information
Set Membership: |
|

PDCD2L
Genomic Map