Assay Details
| Gene Symbol: FXYD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692320.3 | XP_005692377.2 | 2 - 3 | 311 | 71 | |
| XM_018063387.1 | XP_017918876.1 | 2 - 3 | 248 | 71 | ||
| XM_018063388.1 | XP_017918877.1 | 2 - 3 | 211 | 71 | ||
| XM_018063389.1 | XP_017918878.1 | 2 - 3 | 250 | 71 | ||
| XM_018063390.1 | XP_017918879.1 | 2 - 3 | 251 | 71 | ||
| GenBank mRNA | JO580536.1 | - | 6 - 7 | 736 | 71 | |
More Information
Set Membership: |
|

FXYD5
Genomic Map