Assay Details
Gene Symbol: ZNF404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692630.3 | XP_005692687.3 | 4 - 5 | 442 | 117 | |
XM_013971415.2 | XP_013826869.2 | 5 - 6 | 569 | 117 | ||
XM_013971416.2 | XP_013826870.2 | 5 - 6 | 485 | 117 | ||
XM_018062544.1 | XP_017918033.1 | 6 - 7 | 737 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
