Assay Details
| Gene Symbol: PSPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697752.3 | XP_005697809.1 | 5 - 6 | 871 | 70 | |
| XM_005697753.3 | XP_005697810.1 | 5 - 6 | 761 | 70 | ||
| GenBank mRNA | JO599376.2 | - | 1 - 2 | 489 | 70 | |
More Information
Set Membership: |
|

PSPH
Genomic Map