Assay Details
| Gene Symbol: LOC108633221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044427.1 | XP_017899916.1 | 3 - 4 | 441 | 94 | |
| GenBank mRNA | JO367983.2 | - | 2 - 3 | 240 | 94 | |
More Information
Set Membership: |
|

LOC108633221
Genomic Map