Assay Details
| Gene Symbol: STK32B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049721.1 | XP_017905210.1 | 8 - 9 | 1113 | 56 | |
| GenBank mRNA | JO436798.2 | - | 4 - 5 | 508 | 56 | |
More Information
Set Membership: |
|

STK32B
Genomic Map