Assay Details
| Gene Symbol: BICD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052494.1 | XP_017907983.1 | 6 - 7 | 2320 | 61 | |
| XM_018052495.1 | XP_017907984.1 | 6 - 7 | 2320 | 61 | ||
| GenBank mRNA | KA342557.1 | - | 6 - 7 | 2291 | 61 | |
More Information
Set Membership: |
|

BICD2
Genomic Map