Assay Details
| Gene Symbol: CHMP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054631.1 | XP_017910120.1 | 5 - 6 | 892 | 71 | |
| GenBank mRNA | KA336498.1 | - | 1 - 2 | 413 | 71 | |
More Information
Set Membership: |
|

CHMP4A
Genomic Map