Assay Details
| Gene Symbol: CLYBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056396.1 | XP_017911885.1 | 6 - 7 | 831 | 62 | |
| XM_018056397.1 | XP_017911886.1 | 6 - 7 | 831 | 62 | ||
| XM_018056398.1 | XP_017911887.1 | 6 - 7 | 831 | 62 | ||
| XM_018056399.1 | XP_017911888.1 | 6 - 7 | 831 | 62 | ||
| XM_018056400.1 | XP_017911889.1 | 5 - 6 | 635 | 62 | ||
More Information
Set Membership: |
|

CLYBL
Genomic Map