Assay Details
| Gene Symbol: C14H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013969044.2 | XP_013824498.1 | 3 - 4 | 380 | 94 | |
| GenBank mRNA | JO587245.2 | - | 2 - 3 | 192 | 94 | |
More Information
Set Membership: |
|

C14H8orf33
Genomic Map