Assay Details
| Gene Symbol: ZFP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059035.1 | XP_017914524.1 | 4 - 5 | 615 | 96 | |
| GenBank mRNA | JO430053.2 | - | 4 - 5 | 426 | 96 | |
| JO632649.2 | - | 4 - 5 | 769 | 96 | ||
More Information
Set Membership: |
|

ZFP91
Genomic Map