Assay Details
| Gene Symbol: LOC102173489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060590.1 | XP_017916079.1 | 9 | 9148 | 70 | |
| XM_018060591.1 | XP_017916080.1 | 3 | 7849 | 70 | ||
More Information
Set Membership: |
|

LOC102173489
Genomic Map