Assay Details
| Gene Symbol: SARM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064294.1 | XP_017919783.1 | 1 - 2 | 844 | 63 | |
| GenBank mRNA | JO577566.1 | - | 8 - 9 | 4130 | 63 | |
More Information
Set Membership: |
|

SARM1
Genomic Map