Assay Details
| Gene Symbol: USH1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063847.1 | XP_017919336.1 | 2 - 3 | 1570 | 69 | |
| GenBank mRNA | JO625246.2 | - | 1 - 2 | 161 | 69 | |
More Information
Set Membership: |
|

USH1G
Genomic Map