Assay Details
| Gene Symbol: LOC102183271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066587.1 | XP_017922076.1 | 8 - 9 | 1660 | 61 | |
| XM_018066589.1 | XP_017922078.1 | 7 - 8 | 1556 | 61 | ||
| XM_018066591.1 | XP_017922080.1 | 8 - 9 | 1661 | 61 | ||
| GenBank mRNA | JO631944.2 | - | 8 - 9 | 1715 | 61 | |
More Information
Set Membership: |
|

LOC102183271
Genomic Map