Assay Details
| Gene Symbol: LOC102183271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066587.1 | XP_017922076.1 | 9 - 10 | 1740 | 59 | |
| XM_018066589.1 | XP_017922078.1 | 8 - 9 | 1636 | 59 | ||
| XM_018066590.1 | XP_017922079.1 | 8 - 9 | 1704 | 59 | ||
| XM_018066591.1 | XP_017922080.1 | 9 - 10 | 1741 | 59 | ||
| GenBank mRNA | JO631944.2 | - | 9 - 10 | 1795 | 59 | |
More Information
Set Membership: |
|

LOC102183271
Genomic Map