Assay Details
| Gene Symbol: MYRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038374.1 | XP_017893863.1 | 6 - 7 | 1012 | 62 | |
| XM_018038375.1 | XP_017893864.1 | 6 - 7 | 1012 | 62 | ||
| GenBank mRNA | JO639847.2 | - | 11 - 12 | 2322 | 62 | |
More Information
Set Membership: |
|

MYRIP
Genomic Map