Assay Details
| Gene Symbol: LOC102183842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040917.1 | XP_017896406.1 | 11 - 12 | 4493 | 69 | |
| XM_018040918.1 | XP_017896407.1 | 10 - 11 | 4348 | 69 | ||
More Information
Set Membership: |
|

LOC102183842
Genomic Map