Assay Details
| Gene Symbol: LOC102183842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040917.1 | XP_017896406.1 | 7 - 8 | 3881 | 83 | |
| XM_018040918.1 | XP_017896407.1 | 7 - 8 | 3880 | 83 | ||
More Information
Set Membership: |
|

LOC102183842
Genomic Map