Assay Details
| Gene Symbol: ATF7IP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974985.2 | XP_013830439.2 | 9 - 10 | 1943 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATF7IP2| Gene Symbol: ATF7IP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974985.2 | XP_013830439.2 | 9 - 10 | 1943 | 76 | |
Set Membership: |
|