Assay Details
| Gene Symbol: CCDC88B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043538.1 | XP_017899027.1 | 7 - 8 | 925 | 72 | |
| GenBank mRNA | JO580626.1 | - | 7 - 8 | 790 | 72 | |
More Information
Set Membership: |
|

CCDC88B
Genomic Map