Assay Details
| Gene Symbol: DEAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042725.1 | XP_017898214.1 | 6 - 7 | 977 | 61 | |
| GenBank mRNA | JO431385.2 | - | 6 - 7 | 825 | 61 | |
More Information
Set Membership: |
|

DEAF1
Genomic Map