Assay Details
| Gene Symbol: CCNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976482.2 | XP_013831936.2 | 11 - 12 | 3954 | 63 | |
| XM_013976483.2 | XP_013831937.2 | 11 - 12 | 4069 | 63 | ||
| XM_018043973.1 | XP_017899462.1 | 11 - 12 | 4066 | 63 | ||
| XM_018043974.1 | XP_017899463.1 | 10 - 11 | 3976 | 63 | ||
| GenBank mRNA | JO587699.2 | - | 6 - 7 | 765 | 63 | |
| KA318227.1 | - | 1 - 2 | 136 | 63 | ||
More Information
Set Membership: |
|

CCNB3
Genomic Map