Assay Details
| Gene Symbol: GPR156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043285.1 | XP_017898774.1 | 6 - 7 | 1048 | 65 | |
| GenBank mRNA | JO406197.2 | - | 3 - 4 | 1064 | 65 | |
| KA346040.1 | - | 3 - 4 | 266 | 65 | ||
More Information
Set Membership: |
|

GPR156
Genomic Map