Assay Details
| Gene Symbol: MRPL44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058904.1 | XP_017914393.1 | 3 - 4 | 885 | 92 | |
| GenBank mRNA | JO623256.2 | - | 3 - 4 | 876 | 92 | |
More Information
Set Membership: |
|

MRPL44
Genomic Map