Assay Details
| Gene Symbol: PPM1J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677874.3 | XP_005677931.2 | 6 - 7 | 1304 | 63 | |
| XM_013962527.2 | XP_013817981.1 | 6 - 7 | 1306 | 63 | ||
| GenBank mRNA | JO627046.2 | - | 4 - 5 | 442 | 63 | |
More Information
Set Membership: |
|

PPM1J
Genomic Map