Assay Details
| Gene Symbol: WASHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680525.3 | XP_005680582.1 | 3 - 4 | 380 | 95 | |
| XM_005680526.3 | XP_005680583.1 | 3 - 4 | 380 | 95 | ||
| GenBank mRNA | JO628235.2 | - | 5 - 6 | 402 | 95 | |
More Information
Set Membership: |
|

WASHC3
Genomic Map