Assay Details
| Gene Symbol: PACSIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048865.1 | XP_017904354.1 | 6 - 7 | 1004 | 58 | |
| XM_018048866.1 | XP_017904355.1 | 6 - 7 | 1006 | 58 | ||
| GenBank mRNA | JO613094.2 | - | 4 - 5 | 2029 | 58 | |
| JO613095.2 | - | 5 - 6 | 2152 | 58 | ||
More Information
Set Membership: |
|

PACSIN2
Genomic Map