Assay Details
| Gene Symbol: ZFP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052611.1 | XP_017908100.1 | 2 - 3 | 282 | 102 | |
| XM_018052612.1 | XP_017908101.1 | 2 - 3 | 275 | 102 | ||
| XM_018052613.1 | XP_017908102.1 | 2 - 3 | 275 | 102 | ||
More Information
Set Membership: |
|

ZFP37
Genomic Map