Assay Details
| Gene Symbol: HPCAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055663.1 | XP_017911152.1 | 3 - 4 | 786 | 61 | |
| XM_018055664.1 | XP_017911153.1 | 3 - 4 | 551 | 61 | ||
| XM_018055665.1 | XP_017911154.1 | 3 - 4 | 620 | 61 | ||
More Information
Set Membership: |
|

HPCAL1
Genomic Map