Assay Details
| Gene Symbol: ACTR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057859.1 | XP_017913348.1 | 8 - 9 | 1732 | 62 | |
| XM_018057860.1 | XP_017913349.1 | 8 - 9 | 1732 | 62 | ||
| GenBank mRNA | JO593913.2 | - | 2 - 3 | 1951 | 62 | |
More Information
Set Membership: |
|

ACTR5
Genomic Map