Assay Details
| Gene Symbol: IMPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689279.3 | XP_005689336.1 | 5 - 6 | 458 | 75 | |
| XM_018058158.1 | XP_017913647.1 | 5 - 6 | 454 | 75 | ||
| GenBank mRNA | JO434854.2 | - | 4 - 5 | 838 | 75 | |
More Information
Set Membership: |
|

IMPA1
Genomic Map