Assay Details
| Gene Symbol: LOC102175315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058173.1 | XP_017913662.1 | 2 - 3 | 294 | 87 | |
| GenBank mRNA | JO428430.1 | - | 2 - 3 | 483 | 87 | |
| JO428442.1 | - | 2 - 3 | 483 | 87 | ||
More Information
Set Membership: |
|

LOC102175315
Genomic Map