Assay Details
| Gene Symbol: LOC102175315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058173.1 | XP_017913662.1 | 4 - 5 | 509 | 127 | |
| XM_018058174.1 | XP_017913663.1 | 3 - 4 | 396 | 127 | ||
| GenBank mRNA | JO428430.1 | - | 4 - 5 | 698 | 127 | |
| JO428442.1 | - | 4 - 5 | 698 | 127 | ||
More Information
Set Membership: |
|

LOC102175315
Genomic Map