Assay Details
| Gene Symbol: TENT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061948.1 | XP_017917437.1 | 4 - 5 | 965 | 113 | |
| XM_018061949.1 | XP_017917438.1 | 4 - 5 | 965 | 113 | ||
| XM_018061950.1 | XP_017917439.1 | 4 - 5 | 965 | 113 | ||
| GenBank mRNA | JT702242.1 | - | 3 - 4 | 296 | 113 | |
| KA313432.1 | - | 4 - 5 | 679 | 113 | ||
More Information
Set Membership: |
|

TENT4B
Genomic Map