Assay Details
| Gene Symbol: TENT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061948.1 | XP_017917437.1 | 8 - 9 | 1542 | 95 | |
| XM_018061949.1 | XP_017917438.1 | 8 - 9 | 1542 | 95 | ||
| XM_018061950.1 | XP_017917439.1 | 8 - 9 | 1542 | 95 | ||
| GenBank mRNA | KA313432.1 | - | 8 - 9 | 1256 | 95 | |
More Information
Set Membership: |
|

TENT4B
Genomic Map