Assay Details
| Gene Symbol: TENT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061948.1 | XP_017917437.1 | 9 - 10 | 1647 | 112 | |
| XM_018061949.1 | XP_017917438.1 | 9 - 10 | 1647 | 112 | ||
| GenBank mRNA | JT702242.1 | - | 8 - 9 | 980 | 112 | |
| KA313432.1 | - | 9 - 10 | 1361 | 112 | ||
More Information
Set Membership: |
|

TENT4B
Genomic Map