Assay Details
| Gene Symbol: LOC102175340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038481.1 | XP_017893970.1 | 4 - 5 | 1022 | 94 | |
| GenBank mRNA | JT684325.1 | - | 2 - 3 | 365 | 94 | |
More Information
Set Membership: |
|

LOC102175340
Genomic Map