Assay Details
| Gene Symbol: TWNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698333.3 | XP_005698390.1 | 1 - 2 | 1817 | 94 | |
| GenBank mRNA | JO425806.1 | - | 4 - 5 | 1738 | 94 | |
| JO585257.2 | - | 4 - 5 | 1696 | 94 | ||
More Information
Set Membership: |
|

TWNK
Genomic Map