Assay Details
| Gene Symbol: NSDHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700492.3 | XP_005700549.2 | 5 - 6 | 698 | 83 | |
| XM_005700493.3 | XP_005700550.2 | 5 - 6 | 679 | 83 | ||
| XM_005700494.3 | XP_005700551.2 | 6 - 7 | 715 | 83 | ||
| XM_018044710.1 | XP_017900199.1 | 6 - 7 | 735 | 83 | ||
| GenBank mRNA | JO409442.2 | - | 3 - 4 | 1559 | 83 | |
| JO429008.2 | - | 5 - 6 | 735 | 83 | ||
More Information
Set Membership: |
|

NSDHL
Genomic Map