Assay Details
| Gene Symbol: BIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047751.1 | XP_017903240.1 | 10 - 11 | 1534 | 96 | |
| XM_018047752.1 | XP_017903241.1 | 10 - 11 | 1531 | 96 | ||
| GenBank mRNA | JO340905.2 | - | 3 - 4 | 630 | 96 | |
More Information
Set Membership: |
|

BIN2
Genomic Map