Assay Details
| Gene Symbol: WASHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048642.1 | XP_017904131.1 | 3 - 4 | 514 | 96 | |
| GenBank mRNA | JO344048.2 | - | 3 - 4 | 368 | 96 | |
| JO643210.2 | - | 3 - 4 | 471 | 96 | ||
More Information
Set Membership: |
|

WASHC1
Genomic Map