Assay Details
| Gene Symbol: GIPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051175.1 | XP_017906664.1 | 9 - 10 | 1205 | 64 | |
| XM_018051176.1 | XP_017906665.1 | 10 - 11 | 1243 | 64 | ||
| XM_018051177.1 | XP_017906666.1 | 8 - 9 | 1109 | 64 | ||
| XM_018051178.1 | XP_017906667.1 | 8 - 9 | 1120 | 64 | ||
| XM_018051179.1 | XP_017906668.1 | 7 - 8 | 1024 | 64 | ||
More Information
Set Membership: |
|

GIPC1
Genomic Map