Assay Details
| Gene Symbol: LOC102189434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965733.2 | XP_013821187.2 | 3 - 4 | 472 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102189434| Gene Symbol: LOC102189434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965733.2 | XP_013821187.2 | 3 - 4 | 472 | 102 | |
Set Membership: |
|