Assay Details
| Gene Symbol: MAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054820.1 | XP_017910309.1 | 2 - 3 | 378 | 56 | |
| GenBank mRNA | JO632488.2 | - | 2 - 3 | 223 | 56 | |
More Information
Set Membership: |
|

MAL
Genomic Map