Assay Details
| Gene Symbol: LOC102172271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054821.1 | XP_017910310.1 | 3 - 4 | 451 | 80 | |
| XM_018054823.1 | XP_017910312.1 | 2 - 3 | 285 | 80 | ||
More Information
Set Membership: |
|

LOC102172271
Genomic Map