Assay Details
| Gene Symbol: HOXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038766.1 | XP_017894255.1 | 1 - 2 | 740 | 67 | |
| GenBank mRNA | JT677391.1 | - | 1 - 2 | 812 | 67 | |
More Information
Set Membership: |
|

HOXD1
Genomic Map