Assay Details
| Gene Symbol: KIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056964.1 | XP_017912453.1 | 12 - 13 | 1166 | 87 | |
| GenBank mRNA | JT700475.1 | - | 12 - 13 | 1153 | 87 | |
| JT720334.1 | - | 10 - 11 | 960 | 87 | ||
More Information
Set Membership: |
|

KIN
Genomic Map