Assay Details
| Gene Symbol: ASXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057698.1 | XP_017913187.1 | 8 - 9 | 774 | 64 | |
| XM_018057699.1 | XP_017913188.1 | 8 - 9 | 771 | 64 | ||
| XM_018057700.1 | XP_017913189.1 | 7 - 8 | 771 | 64 | ||
| XM_018057701.1 | XP_017913190.1 | 8 - 9 | 681 | 64 | ||
| XM_018057702.1 | XP_017913191.1 | 8 - 9 | 682 | 64 | ||
| XM_018057703.1 | XP_017913192.1 | 6 - 7 | 663 | 64 | ||
More Information
Set Membership: |
|

ASXL1
Genomic Map